Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101142.1 | NP_001094612.1 | 2 - 3 | 733 | 69 | |
GenBank mRNA | BC151545.1 | - | 2 - 3 | 733 | 69 | |
BT026190.1 | - | 2 - 3 | 729 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |