Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128501.2 | NP_001121973.1 | 4 - 5 | 426 | 93 | |
GenBank mRNA | BC103020.1 | - | 4 - 5 | 427 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |