Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128501.2 | NP_001121973.1 | 5 - 6 | 642 | 70 | |
GenBank mRNA | BC103020.1 | - | 5 - 6 | 643 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |