Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130746.2 | NP_001124218.1 | 2 - 3 | 699 | 88 | |
GenBank mRNA | BC120269.1 | - | 2 - 3 | 700 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |