Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130746.2 | NP_001124218.1 | 5 - 6 | 1247 | 70 | |
GenBank mRNA | BC120269.1 | - | 5 - 6 | 1248 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |