Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130751.1 | NP_001124223.1 | 3 - 4 | 319 | 71 | |
GenBank mRNA | BC105544.1 | - | 3 - 4 | 319 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |