Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIMAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079769.1 | NP_001073237.1 | 1 - 2 | 125 | 94 | |
GenBank mRNA | BC103445.1 | - | 1 - 2 | 125 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |