Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082451.3 | NP_001075920.2 | 17 - 18 | 2829 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |