Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082451.3 | NP_001075920.2 | 3 - 4 | 566 | 65 | |
GenBank mRNA | BC133514.1 | - | 3 - 4 | 623 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |