Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103221.1 | NP_001096691.1 | 2 - 3 | 516 | 76 | |
GenBank mRNA | BC134456.1 | - | 2 - 3 | 516 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |