Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105497.2 | NP_001098967.1 | 4 - 5 | 405 | 71 | |
GenBank mRNA | BC116154.1 | - | 4 - 5 | 406 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |