Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159818.1 | NP_001153290.1 | 1 - 2 | 177 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |