Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163244.2 | NP_001156716.1 | 4 - 5 | 1136 | 87 | |
GenBank mRNA | DT829759.1 | - | 5 - 6 | 424 | 87 | |
HX928062.1 | - | 4 - 5 | 405 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |