Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163414.1 | NP_001156886.1 | 4 - 5 | 256 | 102 | |
GenBank mRNA | GQ204112.1 | - | 4 - 5 | 150 | 102 | |
GQ204113.1 | - | 4 - 5 | 256 | 102 | ||
GQ204115.1 | - | 4 - 5 | 143 | 102 | ||
GQ204116.1 | - | 4 - 5 | 143 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Bos taurus Probe spans exons |