Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163786.1 | NP_001157258.1 | 11 - 12 | 1443 | 116 | |
GenBank mRNA | BC123560.1 | - | 11 - 12 | 1443 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |