Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163920.1 | NP_001157392.1 | 6 - 7 | 1368 | 87 | |
GenBank mRNA | BT030550.1 | - | 6 - 7 | 1368 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |