Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205510.3 | NP_001192439.1 | 2 - 3 | 660 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |