Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205523.2 | NP_001192452.2 | 3 - 4 | 272 | 114 | |
GenBank mRNA | BC111307.1 | - | 3 - 4 | 273 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |