Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205523.2 | NP_001192452.2 | 7 - 8 | 677 | 81 | |
GenBank mRNA | BC111307.1 | - | 7 - 8 | 678 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |