Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205549.1 | NP_001192478.1 | 8 - 9 | 1897 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |