Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205604.1 | NP_001192533.1 | 6 - 7 | 891 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |