Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205658.1 | NP_001192587.1 | 6 - 7 | 1680 | 63 | |
GenBank mRNA | BC149001.1 | - | 7 - 8 | 2237 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |