Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205688.1 | NP_001192617.1 | 3 - 4 | 594 | 73 | |
GenBank mRNA | EU616677.1 | - | 3 - 4 | 421 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |