Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205718.2 | NP_001192647.2 | 3 - 4 | 510 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |