Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205728.1 | NP_001192657.1 | 7 - 8 | 4250 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |