Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205748.2 | NP_001192677.1 | 6 - 7 | 2854 | 80 | |
GenBank mRNA | BC149148.1 | - | 2 - 3 | 1022 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |