Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205764.1 | NP_001192693.1 | 6 - 7 | 869 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |