Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205766.3 | NP_001192695.1 | 1 - 2 | 807 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Bos taurus Probe spans exons |