Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205771.1 | NP_001192700.1 | 8 - 9 | 1795 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Bos taurus Probe spans exons |