Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205809.1 | NP_001192738.1 | 6 - 7 | 1279 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |