Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205837.1 | NP_001192766.1 | 4 - 5 | 1832 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |