Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205868.1 | NP_001192797.1 | 4 - 5 | 754 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |