Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205869.2 | NP_001192798.1 | 11 - 12 | 1959 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |