Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205869.2 | NP_001192798.1 | 7 - 8 | 1327 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |