Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205888.2 | NP_001192817.1 | 2 - 3 | 400 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |