Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205899.3 | NP_001192828.1 | 11 - 12 | 1573 | 84 | |
NM_001317187.2 | NP_001304116.1 | 11 - 12 | 1573 | 84 | ||
GenBank mRNA | EE347213.1 | - | 3 - 4 | 329 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |