Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205912.1 | NP_001192841.1 | 9 - 10 | 1302 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |