Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTPAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205931.2 | NP_001192860.1 | 4 - 5 | 854 | 111 | |
GenBank mRNA | BC149824.1 | - | 3 - 4 | 744 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |