Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205963.2 | NP_001192892.2 | 5 - 6 | 859 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |