Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205984.3 | NP_001192913.3 | 3 - 4 | 468 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |