Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001354198.1 | NP_001341127.1 | 2 - 3 | 736 | 63 | |
GenBank mRNA | EH139359.1 | - | 1 - 2 | 74 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |