Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206021.1 | NP_001192950.1 | 1 - 2 | 708 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Probe spans exons |