Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206034.1 | NP_001192963.1 | 7 - 8 | 1873 | 78 | |
GenBank mRNA | BT030559.1 | - | 3 - 4 | 1409 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |