Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206075.1 | NP_001193004.1 | 4 - 5 | 318 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |