Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3H1orf87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206121.3 | NP_001193050.3 | 10 - 11 | 1332 | 99 | |
GenBank mRNA | BC149431.1 | - | 10 - 11 | 1363 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |