Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206157.1 | NP_001193086.1 | 7 - 8 | 1321 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |