Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206221.1 | NP_001193150.1 | 3 - 4 | 285 | 77 | |
GenBank mRNA | BC103314.1 | - | 3 - 4 | 256 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |