Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206240.1 | NP_001193169.1 | 3 - 4 | 743 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |