Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FA2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192455.1 | NP_001179384.1 | 4 - 5 | 654 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |