Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192462.2 | NP_001179391.2 | 7 - 8 | 1117 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |